Assay Details
| Gene Symbol: NANOS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111100.4 | XP_001111100.3 | 1479 | 102 | ||
| GenBank mRNA | JV059896.1 | - | 636 | 99 | ||
| JV650245.1 | - | 655 | 99 | |||
More Information
Set Membership: |
|

NANOS3
Genomic Map