Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808287.3 | XP_002808333.2 | 857 | 69 | ||
| XM_028840626.1 | XP_028696459.1 | 862 | 69 | |||
| XM_028840627.1 | XP_028696460.1 | 707 | 69 | |||
| GenBank mRNA | JV672677.1 | - | 198 | 69 | ||
More Information
Set Membership: |
|

SEPTIN12
Genomic Map