Assay Details
| Gene Symbol: COL6A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132202.2 | XP_014987688.2 | 92 | |||
| XM_015132203.2 | XP_014987689.2 | 92 | ||||
| XM_015132205.2 | XP_014987691.2 | 92 | ||||
| XM_028843686.1 | XP_028699519.1 | 92 | ||||
| XM_028843687.1 | XP_028699520.1 | 92 | ||||
| GenBank mRNA | JV051921.1 | - | 183 | 92 | ||
More Information
Set Membership: |
|

COL6A6
Genomic Map