Assay Details
| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129771.2 | XP_014985257.1 | 1063 | 75 | ||
| GenBank mRNA | JV615163.1 | - | 625 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTR6| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129771.2 | XP_014985257.1 | 1063 | 75 | ||
| GenBank mRNA | JV615163.1 | - | 625 | 75 | ||
Set Membership: |
|