Assay Details
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116338.2 | XP_014971824.1 | 714 | 96 | ||
| GenBank mRNA | JU321056.1 | - | 676 | 96 | ||
| JU472284.1 | - | 688 | 96 | |||
| JV045140.1 | - | 624 | 96 | |||
| JV600475.1 | - | 560 | 96 | |||
More Information
Set Membership: |
|

PHPT1
Genomic Map