Assay Details
| Gene Symbol: RNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150141.2 | XP_015005627.1 | 1107 | 67 | ||
| XM_015150143.2 | XP_015005629.1 | 1106 | 67 | |||
| GenBank mRNA | JU326948.1 | - | 850 | 67 | ||
| JV644679.1 | - | 853 | 67 | |||
| JV708956.1 | - | 887 | 67 | |||
More Information
Set Membership: |
|

RNF215
Genomic Map