Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146428.2 | XP_015001914.2 | 636 | 74 | ||
| GenBank mRNA | JU330656.1 | - | 407 | 74 | ||
| JU494588.1 | - | 420 | 74 | |||
| JV663009.1 | - | 407 | 74 | |||
More Information
Set Membership: |
|

TAF3
Genomic Map