Assay Details
| Gene Symbol: ZFP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118378.2 | XP_014973864.1 | 319 | 71 | ||
| GenBank mRNA | JU332898.1 | - | 98 | 71 | ||
| JU474288.1 | - | 72 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015118378.2 transcripts.
Set Membership: |
|

ZFP3
Genomic Map