Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121710.2 | XP_014977196.2 | 3217 | 119 | ||
| XM_028837864.1 | XP_028693697.1 | 1462 | 119 | |||
| XM_028837865.1 | XP_028693698.1 | 1825 | 119 | |||
| XM_028837866.1 | XP_028693699.1 | 1639 | 119 | |||
| XM_028837867.1 | XP_028693700.1 | 1292 | 119 | |||
| XM_028837869.1 | XP_028693702.1 | 1602 | 119 | |||
| GenBank mRNA | JV712613.1 | - | 215 | 120 | ||
More Information
Set Membership: |
|

KCTD1
Genomic Map