Assay Details
| Gene Symbol: LOC693354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU323531.1 | - | 71 | 64 | ||
| JU481634.1 | - | 104 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC693354| Gene Symbol: LOC693354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU323531.1 | - | 71 | 64 | ||
| JU481634.1 | - | 104 | 64 | |||
Set Membership: |
|