Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141427.2 | XP_014996913.2 | 1003 | 111 | ||
| GenBank mRNA | JU321010.1 | - | 514 | 111 | ||
| JV044548.1 | - | 532 | 111 | |||
More Information
Set Membership: |
|

BOD1
Genomic Map