Assay Details
| Gene Symbol: RRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111692.2 | XP_014967178.1 | 659 | 86 | ||
| XM_028831975.1 | XP_028687808.1 | 897 | 86 | |||
| GenBank mRNA | JU491200.1 | - | 600 | 86 | ||
| JV068095.1 | - | 591 | 86 | |||
More Information
Set Membership: |
|

RRM2
Genomic Map