Assay Details
| Gene Symbol: PRKAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140450.2 | XP_014995936.1 | 175 | 97 | ||
| GenBank mRNA | JU328227.1 | - | 86 | 97 | ||
| JV655617.1 | - | 92 | 97 | |||
More Information
Set Membership: |
|

PRKAA2
Genomic Map