Assay Details
Gene Symbol: LOC694533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841707.1 | XP_028697540.1 | 2235 | 94 | ||
XM_028841715.1 | XP_028697548.1 | 2372 | 94 | |||
XM_028841716.1 | XP_028697549.1 | 3863 | 94 | |||
GenBank mRNA | JV049093.1 | - | 617 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
