Assay Details
| Gene Symbol: PSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101767.4 | XP_001101767.3 | 1072 | 119 | ||
| XM_015117956.2 | XP_014973442.1 | 1731 | 119 | |||
| XM_028835269.1 | XP_028691102.1 | 748 | 119 | |||
| GenBank mRNA | JU334226.1 | - | 763 | 119 | ||
| JU475340.1 | - | 804 | 119 | |||
| JV046889.1 | - | 808 | 119 | |||
| JV682003.1 | - | 804 | 119 | |||
More Information
Set Membership: |
|

PSAT1
Genomic Map