Assay Details
| Gene Symbol: HCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839480.1 | XP_028695313.1 | 1273 | 80 | ||
| GenBank mRNA | JU346815.1 | - | 785 | 80 | ||
| JV589999.1 | - | 808 | 80 | |||
| JV692570.1 | - | 811 | 80 | |||
More Information
Set Membership: |
|

HCN2
Genomic Map