Assay Details
| Gene Symbol: FNTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145263.2 | XP_015000749.1 | 835 | 68 | ||
| XM_028852645.1 | XP_028708478.1 | 835 | 68 | |||
| GenBank mRNA | JU327081.1 | - | 191 | 68 | ||
| JV046583.1 | - | 209 | 68 | |||
| JV678315.1 | - | 236 | 68 | |||
More Information
Set Membership: |
|

FNTA
Genomic Map