Assay Details
| Gene Symbol: TOMM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117420.2 | XP_014972906.2 | 507 | 96 | ||
| GenBank mRNA | JU328058.1 | - | 312 | 96 | ||
| JV047048.1 | - | 475 | 96 | |||
| JV638454.1 | - | 485 | 96 | |||
More Information
Set Membership: |
|

TOMM5
Genomic Map