Assay Details
| Gene Symbol: POLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133645.2 | XP_014989131.2 | 275 | 72 | ||
| XM_028845843.1 | XP_028701676.1 | 288 | 72 | |||
| XM_028845844.1 | XP_028701677.1 | 290 | 72 | |||
| GenBank mRNA | JU484624.1 | - | 118 | 72 | ||
More Information
Set Membership: |
|

POLM
Genomic Map