Assay Details
| Gene Symbol: TRMT10C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129663.2 | XP_014985149.1 | 353 | 107 | ||
| GenBank mRNA | JU488860.1 | - | 321 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRMT10C| Gene Symbol: TRMT10C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129663.2 | XP_014985149.1 | 353 | 107 | ||
| GenBank mRNA | JU488860.1 | - | 321 | 107 | ||
Set Membership: |
|