Assay Details
| Gene Symbol: MED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141317.2 | XP_014996803.1 | 753 | 87 | ||
| GenBank mRNA | JU337178.1 | - | 715 | 87 | ||
| JU477906.1 | - | 682 | 87 | |||
More Information
Set Membership: |
|

MED7
Genomic Map