Assay Details
| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV075440.1 | - | 621 | 56 | ||
| JV075441.1 | - | 605 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALML4| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV075440.1 | - | 621 | 56 | ||
| JV075441.1 | - | 605 | 56 | |||
Set Membership: |
|