Assay Details
| Gene Symbol: PRORP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085026.4 | XP_001085026.3 | 462 | 110 | ||
| XM_001085149.4 | XP_001085149.2 | 356 | 110 | |||
| XM_015143563.2 | XP_014999049.2 | 457 | 110 | |||
| XM_015143564.2 | XP_014999050.2 | 425 | 110 | |||
| GenBank mRNA | JU351074.1 | - | 201 | 110 | ||
| JV727673.1 | - | 218 | 110 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001085026.4, XM_001085149.4, XM_015143563.2, XM_015143564.2 transcripts.
Set Membership: |
|

PRORP
Genomic Map