Assay Details
| Gene Symbol: TXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150236.2 | XP_015005722.1 | 756 | 76 | ||
| XM_015150238.2 | XP_015005724.1 | 638 | 76 | |||
| GenBank mRNA | JU326253.1 | - | 117 | 76 | ||
| JU473495.1 | - | 184 | 76 | |||
| JV045066.1 | - | 236 | 76 | |||
| JV566168.1 | - | 219 | 76 | |||
More Information
Set Membership: |
|

TXN2
Genomic Map