Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149175.2 | XP_015004661.1 | 1785 | 96 | ||
| XM_015149177.2 | XP_015004663.1 | 1843 | 96 | |||
| XM_028828337.1 | XP_028684170.1 | 1534 | 96 | |||
| XM_028828338.1 | XP_028684171.1 | 1599 | 96 | |||
| GenBank mRNA | JU321436.1 | - | 1619 | 96 | ||
| JU471194.1 | - | 1646 | 96 | |||
| JV044665.1 | - | 1620 | 96 | |||
| JV565420.1 | - | 1621 | 96 | |||
More Information
Set Membership: |
|

AHCY
Genomic Map