Assay Details
| Gene Symbol: LY9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846611.1 | XP_028702444.1 | 1322 | 57 | ||
| GenBank mRNA | JU486669.1 | - | 1219 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY9| Gene Symbol: LY9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846611.1 | XP_028702444.1 | 1322 | 57 | ||
| GenBank mRNA | JU486669.1 | - | 1219 | 57 | ||
Set Membership: |
|