Assay Details
| Gene Symbol: STPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802234.3 | XP_002802280.2 | 1335 | 113 | ||
| XM_015131276.2 | XP_014986762.2 | 1234 | 113 | |||
| XM_015131284.2 | XP_014986770.1 | 1079 | 113 | |||
| GenBank mRNA | JV075048.1 | - | 324 | 113 | ||
More Information
Set Membership: |
|

STPG1
Genomic Map