Assay Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098517.4 | XP_001098517.1 | 368 | 63 | ||
XM_015134924.2 | XP_014990410.1 | 551 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
