Assay Details
| Gene Symbol: FBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091930.3 | XP_001091930.1 | 666 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001091930.3 transcripts.
Set Membership: |
|

FBLL1
Genomic Map