Assay Details
| Gene Symbol: LOC701312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121551.2 | XP_014977037.1 | 954 | 96 | ||
| Gene Symbol: MC2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837949.1 | XP_028693782.1 | 954 | 96 | ||
More Information
Set Membership: |
|

LOC701312
Genomic Map