Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128550.2 | XP_014984036.2 | 635 | 64 | ||
| Gene Symbol: OPN1MW3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842323.1 | XP_028698156.1 | 64 | |||
More Information
Set Membership: |
|

OPN1LW
Genomic Map