Assay Details
| Gene Symbol: LOC694004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846016.1 | XP_028701849.1 | 4594 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC694004| Gene Symbol: LOC694004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846016.1 | XP_028701849.1 | 4594 | 86 | ||