Assay Details
| Gene Symbol: LOC694004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846016.1 | XP_028701849.1 | 258 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028846016.1 transcripts.
Set Membership: |
|

LOC694004
Genomic Map