Assay Details
Gene Symbol: SLC49A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848201.1 | XP_028704034.1 | 1202 | 118 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
