Assay Details
| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836172.1 | XP_028692005.1 | 5570 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM35B| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836172.1 | XP_028692005.1 | 5570 | 93 | ||
Set Membership: |
|