Assay Details
| Gene Symbol: PTTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138200.2 | XP_014993686.1 | 2121 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015138200.2 transcripts.
Set Membership: |
|

PTTG2
Genomic Map