Assay Details
| Gene Symbol: AMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851888.1 | XP_028707721.1 | 357 | 77 | ||
| XM_028851889.1 | XP_028707722.1 | 357 | 77 | |||
| XM_028851890.1 | XP_028707723.1 | 357 | 77 | |||
| XM_028851891.1 | XP_028707724.1 | 357 | 77 | |||
| XM_028851892.1 | XP_028707725.1 | 1214 | 77 | |||
| XM_028851893.1 | XP_028707726.1 | 357 | 77 | |||
| XM_028851894.1 | XP_028707727.1 | 357 | 77 | |||
| XM_028851895.1 | XP_028707728.1 | 357 | 77 | |||
More Information
Set Membership: |
|

AMN
Genomic Map