Assay Details
| Gene Symbol: LOC707623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849926.1 | XP_028705759.1 | 813 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028849926.1 transcripts.
Set Membership: |
|

LOC707623
Genomic Map