Assay Details
| Gene Symbol: LOC699013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127685.2 | XP_014983171.1 | 2013 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC699013| Gene Symbol: LOC699013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127685.2 | XP_014983171.1 | 2013 | 77 | ||
Set Membership: |
|