Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834039.1 | XP_028689872.1 | 286 | 107 | ||
| XM_028834040.1 | XP_028689873.1 | 286 | 107 | |||
| XM_028834041.1 | XP_028689874.1 | 286 | 107 | |||
More Information
Set Membership: |
|

TTC36
Genomic Map