Assay Details
| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130554.2 | XP_014986040.1 | 1950 | 96 | ||
| XM_028838298.1 | XP_028694131.1 | 2009 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTR1D| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130554.2 | XP_014986040.1 | 1950 | 96 | ||
| XM_028838298.1 | XP_028694131.1 | 2009 | 96 | |||
Set Membership: |
|