Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138555.2 | XP_014994041.1 | 802 | 133 | ||
| XM_028848513.1 | XP_028704346.1 | 551 | 133 | |||
More Information
Set Membership: |
|

NPFFR2
Genomic Map