Assay Details
| Gene Symbol: LOC714004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103553.4 | XP_001103553.2 | 513 | 88 | ||
| XM_015144127.2 | XP_014999613.1 | 614 | 88 | |||
| XM_028831953.1 | XP_028687786.1 | 514 | 88 | |||
| XM_028831955.1 | XP_028687788.1 | 514 | 88 | |||

LOC714004
Genomic Map