Assay Details
| Gene Symbol: LOC718794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839252.1 | XP_028695085.1 | 710 | 75 | ||
| XM_028839253.1 | XP_028695086.1 | 1377 | 75 | |||
More Information
Set Membership: |
|

LOC718794
Genomic Map