Assay Details
| Gene Symbol: C15H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117661.1 | XP_014973147.1 | 55 | 124 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H9orf92| Gene Symbol: C15H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117661.1 | XP_014973147.1 | 55 | 124 | ||
Set Membership: |
|