Assay Details
| Gene Symbol: CACNG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120160.2 | XP_014975646.1 | 431 | 71 | ||
| GenBank mRNA | JU326290.1 | - | 205 | 71 | ||
| JV637926.1 | - | 240 | 71 | |||
More Information
Set Membership: |
|

CACNG4
Genomic Map