Assay Details
| Gene Symbol: LOC106997075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131894.2 | XP_014987380.1 | 818 | 183 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131894.2 transcripts.
Set Membership: |
|

LOC106997075
Genomic Map