Assay Details
| Gene Symbol: LOC719436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124675.2 | XP_014980161.2 | 521 | 83 | ||
| XM_015124676.2 | XP_014980162.2 | 607 | 83 | |||
| XM_015124677.2 | XP_014980163.2 | 521 | 83 | |||
| XM_015124678.2 | XP_014980164.2 | 361 | 83 | |||
| XM_028839831.1 | XP_028695664.1 | 357 | 83 | |||
More Information
Set Membership: |
|

LOC719436
Genomic Map