Assay Details
Gene Symbol: SPRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847999.1 | XP_028703832.1 | 216 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028847999.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
