Assay Details
Gene Symbol: MMP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086245.4 | XP_001086245.3 | 136 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
